Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Tris Borate EDTA
SCI_ED

Catalog No. s25616 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
S25616 500 mL
1 options

Catalog No. S25616

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Chemical Name or Material Tris Borate EDTA
CAS 10043-35-3, 6381-92-6, 77-86-1, 7732-18-5
Density ∽1g/mL
Quantity 500 mL
Assay Percent Range 2.75% w/v
Packaging Liquid
Synonym TBE 5X Buffer
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.