Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS-buffered saline (TBS, 10X), with 1% Triton™ X-100, Thermo Scientific Chemicals

Catalog No. p-7042438
Click to view available options
Quantity:
500 mL

It used in some biochemical techniques that is maintain the pH within a relatively narrow range. Tris-HCl is commonly used to make TBS buffers. Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash micro titer plates for western blot experiments and screening methods. it is suitable for use as a washing buffer for alkaline phosphatase or peroxidase conjugates in dot blot assay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It used in some biochemical techniques that is maintain the pH within a relatively narrow range. Tris-HCl is commonly used to make TBS buffers. Tris Buffered Saline has been used as washing buffer for electrodes. It was used to wash micro titer plates for western blot experiments and screening methods. it is suitable for use as a washing buffer for alkaline phosphatase or peroxidase conjugates in dot blot assay.

Solubility
Fully miscible in water.

Notes
250mM Tris, 27mM KCl, 1.37M NaCl, pH 7.4
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00132476
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 10X
Physical Form Liquid
Quantity 500 mL
Solubility Information Fully miscible in water.
Chemical Name or Material TRIS-buffered saline, with 1% Triton™ X-100
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.