Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS-Buffered Saline (TBS, 10X) pH 7.6, PROMO, Thermo Scientific Chemicals

Catalog No. J62662K7PM
Click to view available options
Quantity:
2 L
4 L

Solubility

Miscible with water.

Notes

250mM Tris, 27mM potassium chloride, 1.37M sodium chloride, pH 7.6.Store in a cool place. Incompatible with strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14 g/mol
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 10X
pH 7.6
Physical Form Liquid
Quantity 4 L
Solubility Information Miscible with water.
Chemical Name or Material TRIS-buffered saline

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.