Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(dibenzoylmethanato) Iron 97.0+%, TCI America™
SDP

Catalog No. T168625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T168625G 25 g
T16865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T168625G Supplier TCI America Supplier No. T168625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14405-49-3
Molecular Formula C45H33FeO6
Molecular Weight (g/mol) 725.60
MDL Number MFCD00192557,MFCD14636458
InChI Key JWPHPBLJVLHKOZ-UHFFFAOYSA-K
Synonym tris dibenzoylmethanato iron, iron iii diphenylpropanedionate
PubChem CID 57369570
IUPAC Name iron(3+) tris(3-oxo-1,3-diphenylprop-1-en-1-olate)
SMILES [Fe+3].[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 25 g
Formula Weight 725.60
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(dibenzoylmethanato) Iron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.