Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(dibenzoylmethane)mono(phenanthroline)europium(III)

Catalog No. AA4417577
Click to view available options
Quantity:
0.1 g
0.5 g

Catalog No. AA4417577

Supplier: Thermo Scientific Chemicals 04417577

Only null left
Add to Cart

As an organic red light emitter | CAS: 17904-83-5 | C57H44EuN2O6 | 1004.951 g/mol

As an organic red light emitter

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17904-83-5
Molecular Formula C57H44EuN2O6
Molecular Weight (g/mol) 1004.951
MDL Number MFCD01321202
InChI Key DYKOLWWJTALFFU-RWBKAWJDSA-N
Synonym eu dbm 3 phen, tris dibenzoylmethane mono phenanthroline europium iii, tris dibenzoylmethane phenanthroline europium iii, tris dibenzoylmethane mono 1,10-phenanthroline europium lll, europium iii tris 1,3-diphenyl-1,3-propanedionato mono 1,10-phenanthroline, 2e-3-bis 1e-3-oxo-1,3-diphenylprop-1-en-1-yl oxy europio oxy-1,3-diphenylprop-2-en-1-one; phen
PubChem CID 14205791
IUPAC Name europium;(E)-3-hydroxy-1,3-diphenylprop-2-en-1-one;1,10-phenanthroline
SMILES C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC=C(C=C1)C(=CC(=O)C2=CC=CC=C2)O.C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.[Eu]

Specifications

Melting Point 172°C to 173°C
Quantity 0.1 g
Solubility Information Soluble in dichloromethane, THF
Formula Weight 1001.93
Physical Form Powder
Chemical Name or Material Tris (dibenzoylmethane)mono (phenanthroline)europium (III)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.