Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris-EDTA Buffer Solutions, R&D Systems™
SDP

Catalog No. NB90066730 Shop All Novus Products
Change view
Click to view available options
pH:
8
9
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
NB90066730 8
NB90062085 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB90066730 Supplier R&D Systems Supplier No. NB90066730

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. For recovery of antigens masked by fixation in cross-linking fixatives, such as formalin

  • 10X EDTA buffer for Immunohistochemistry
  • Dilute one part buffer with nine parts distilled water
  • Unit size: 500mL

Recommended Storage

Store at room temperature

Specifications

Chemical Name or Material Tris-EDTA Buffer
pH 8
Product Line Novus
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.