Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(ethylenediamine)cobalt chloride trihydrate, 97%, Thermo Scientific™

Catalog No. p-7077765
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 14883-80-8
Molecular Formula C6H24Cl3CoN6·3H2O
Molecular Weight (g/mol) 399.64
InChI Key WBMVTRUQSMZIKW-UHFFFAOYSA-K
Synonym tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride
PubChem CID 73995044
IUPAC Name cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate
SMILES C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3]

Specifications

Melting Point >300.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 399.64
Percent Purity 97%
Chemical Name or Material Tris(ethylenediamine)cobalt chloride trihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.