Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tris-Glycine, 10X Solution (Electrophoresis), Fisher BioReagents™
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Nalgene Poly Bottle
Poly CUBE
Description
Tris-Glycine is commonly used as a buffer in native polyacrylamide gel electrophoresis.
Specifications
Specifications
| Chemical Name or Material | Tris-Glycine |
| Name Note | 10X Solution |
| Color | Undesignated |
| CAS | 77-86-1, 56-40-6 |
| Physical Form | Liquid |
| Quantity | 1 L |
| DNase | DNase free |
| Filtered Through | Filtered through a 0.2-micron filter. |
| Purity Grade Notes | DNase-, RNase- and Protease-Free |
| Packaging | Nalgene Poly Bottle |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction