Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS Hydrochloride, Made from Tromethamine USP, Biotech Reagent Grade, J.T. Baker™

Catalog No. 02004517 Shop All Avantor Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Melting Point 147°C to 153°C
Packaging Poly Bottle
Quantity 2.5 kg
Specific Gravity 1L = 0.00kg
Formula Weight 157.6
Grade GMP manufactured product
Chemical Name or Material TRIS Hydrochloride, Made from Tromethamine USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.