Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIS Hydrochloride, Ultrapure Bioreagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
Description
- For Liquid Chromatography and Molecular Biology Applications
Specifications
Specifications
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Formula Weight | 157.6 |
| Grade | Ultrapure Bioreagent |
| Chemical Name or Material | TRIS Hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction