Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent

Catalog No. p-209362
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, TRIS, Tris buffer, Tromethamine, Tromethane
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Absorbance 0.2 max. at 290nm, (40% aq. soln.) (1 cm cell), (40% aq. soln.) (1 cm cell) at 0nm
Absorbance Conditions (40% aq. soln.) (1 cm cell)
Melting Point >168.5°C
Boiling Point 219.0°C to 220.0°C (10.0mmHg)
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 100 g
Assay Percent Range 99.8%
Infrared Spectrum Authentic
Linear Formula (HOCH2)3CNH2
Packaging Plastic bottle
Beilstein 04, 303
Merck Index 15, 9951
Formula Weight 121.14
Percent Purity 99.8%
Grade ACS Reagent
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-064-4
RTECSNumber TY2900000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.