Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIS, MP Biomedicals™

Catalog No. p-7083161
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg

TRIS, Molecular Biology Reagent, ≥99.95%, White Crystalline Powder

  • Tris has been useful as buffers in a wide variety of biological systems.
  • Uses include pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.
  • Tris has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes).
  • Tris does not precipitate calcium salts and is of value in maintaining solubility of manganese salts.
  • It can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid.
  • It is an auxiliary material in pharmaceutical science
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 167°C to 172°C
CAS 77-86-1
Boiling Point 219°C to 220°C at 10 mmHg (Literature)
Color White
pH 10 to 11.5 (5% aq. soln.)
Quantity 100 g
Purity Grade Notes ≥99.95%
Loss on Drying ≤1%
Molecular Formula C4H11NO3
MDL Number MFCD00004679
Synonym Trometamol, THAM, Tromethamine, Tris base
Solubility Information Soluble in water (40% aq soln) Clear, Colorless Solution (550mg/mL), ethyleneglycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL).
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Molecular Weight (g/mol) 121.14
Grade Molecular Biology Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.