Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(pentafluorophenyl)phosphine, 97%

Catalog No. p-209702
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 1259-35-4 | C18F15P | 532.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1259-35-4
Molecular Formula C18F15P
Molecular Weight (g/mol) 532.15
MDL Number MFCD00079654
InChI Key FQLSDFNKTNBQLC-UHFFFAOYSA-N
Synonym tris pentafluorophenyl phosphine, phosphine, tris pentafluorophenyl, tris perfluorophenyl phosphine, tris pentafluorophenyl phosphane, tris 2,3,4,5,6-pentafluorophenyl phosphane, pubchem6422, acmc-1bogg, ksc491k1f, tris-pentafluorophenyl-phosphane
PubChem CID 102076
IUPAC Name tris(2,3,4,5,6-pentafluorophenyl)phosphane
SMILES FC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 108.0°C to 110.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6F5)3P
Quantity 1 g
Formula Weight 531.98
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tris(pentafluorophenyl)phosphine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-021-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.