Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(trimethylsilyl) borate, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
4325-85-3
C9H27BO3Si3
278.38
MFCD00051588
YZYKZHPNRDIPFA-UHFFFAOYSA-N
tris trimethylsilyl borate,tris trimethylsiloxy boron,silanol, trimethyl-, triester with boric acid h3bo3,tritrimethylsilyl borate,borane, tris trimethylsiloxy,silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3,acmc-209jtk,tris trimethylsiloxy borane,boric acid, 3tms derivative
78020
tris(trimethylsilyl) borate
B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
4325-85-3
C9H27BO3Si3
278.38
MFCD00051588
YZYKZHPNRDIPFA-UHFFFAOYSA-N
tris trimethylsilyl borate,tris trimethylsiloxy boron,silanol, trimethyl-, triester with boric acid h3bo3,tritrimethylsilyl borate,borane, tris trimethylsiloxy,silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3,acmc-209jtk,tris trimethylsiloxy borane,boric acid, 3tms derivative
78020
tris(trimethylsilyl) borate
B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
Boiling Point 183°C to 185°C
Specific Gravity 0.834
Flash Point 42°C
Density 0.8340g/mL
Refractive Index 1.3850 to 1.387
Color Colorless
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula B[OSi(CH3)3]
Formula Weight 278.38
Physical Form Liquid
Percent Purity ≥97.0%
Chemical Name or Material Tris(trimethylsilyl) borate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning