Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(trimethylsilyl) borate

Catalog No. p-3523624
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 4325-85-3 | C9H27BO3Si3 | 278.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4325-85-3
Molecular Formula C9H27BO3Si3
Molecular Weight (g/mol) 278.38
MDL Number MFCD00051588
InChI Key YZYKZHPNRDIPFA-UHFFFAOYSA-N
Synonym tris trimethylsilyl borate, tris trimethylsiloxy boron, silanol, trimethyl-, triester with boric acid h3bo3, tritrimethylsilyl borate, borane, tris trimethylsiloxy, silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3, acmc-209jtk, tris trimethylsiloxy borane, boric acid, 3tms derivative
PubChem CID 78020
IUPAC Name tris(trimethylsilyl) borate
SMILES B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Density 0.8340g/mL
Boiling Point 183°C to 185°C
Flash Point 42°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3850 to 1.387
Linear Formula B[OSi(CH3)3]
Quantity 25 g
Specific Gravity 0.834
Formula Weight 278.38
Percent Purity ≥97.0%
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyl) borate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.