Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(trimethylsilyl) Phosphate 98.0+%, TCI America™
SDP

Catalog No. P124825G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 10497-05-9
Molecular Formula C9H27O4PSi3
Molecular Weight (g/mol) 314.54
MDL Number MFCD00008267
InChI Key QJMMCGKXBZVAEI-UHFFFAOYSA-N
Synonym Phosphoric Acid Tris(trimethylsilyl) Ester
PubChem CID 25317
IUPAC Name tritrimethylsilyl phosphate
SMILES C[Si](C)(C)OP(=O)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 236°C
Quantity 25 g
UN Number 1993
Formula Weight 314.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyl) Phosphate
RTECSNumber TC9700000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.