Learn More
Tris(trimethylsilyl) phosphite, 95%
Description
Tris(trimethylsilyl) phosphite is a reagent used for the mild preparation of bis(TMS) phosphonates (Arbuzow reaction). It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.893 g/mL |
| Boiling Point | 80°C (8 mmHg) |
| Refractive Index | 1.409 |
| Quantity | 0.5 g |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Sparingly soluble in water. |
| Formula Weight | 298.54 |
| Percent Purity | 96% |
| Physical Form | Liquid |
| Chemical Name or Material | Tris(trimethylsilyl) phosphite |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.