Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trismat(R) Hydrochloride, Biologics Plus, cGMP, bioCERTIFIED™, 2.5 kg, Spectrum Chemical

Click to view available options
Quantity:
2.5 kg
Specifications
Specifications
Chemical Name or Material | Trismat(R) Hydrochloride |
CAS | 1185-53-1 |
CAS Max % | 100% |
For Use With (Application) | Molecular Biology |
pH | 3.7 to 4.7 |
Physical Form | Crystalline Powder |
Quantity | 2.5 kg |
Percent Purity | 99.5% |
Grade | Biologics Plus, cGMP, bioCERTIFIED |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction