Learn More
Trisodium citrate dihydrate, 99%, Thermo Scientific Chemicals
Description
Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents cleaning, furnishing care products, laundry dishwashing products and cleaning automobile radiators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trisodium citrate dihydrate, is widely applied in food, beverages and fillers as a buffering, sequestering or an emulsifying agent. It used as an anticoagulant in blood transfusions, osmotic laxative, functional fluids, solvents cleaning, furnishing care products, laundry dishwashing products and cleaning automobile radiators.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Stable and incompatible with bases, reducing agents, oxidizing agents.
Specifications
Melting Point | 150°C (in -2H2O) |
Density | 1.76 |
Odor | Odorless |
Quantity | 500 g |
Beilstein | 6104939 |
Merck Index | 14,8602 |
Solubility Information | Soluble in water. Insoluble in alcohol. |
Formula Weight | 294.10 (258.07 Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | Trisodium citrate dihydrate |
Safety and Handling
Hazard Category | H500 |
EINECSNumber | 200-675-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.