Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trisodium Naphthalene-1,3,6-trisulfonate Hydrate 65.0+%, TCI America™
SDP

Catalog No. N0021100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0021100G 100 g
N002125G 25 g
N0021500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0021100G Supplier TCI America Supplier No. N0021100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5182-30-9
Molecular Formula C10H5Na3O9S3
Molecular Weight (g/mol) 434.29
MDL Number MFCD00003989
InChI Key NJPKYOIXTSGVAN-UHFFFAOYSA-K
Synonym trisodium 1,3,6-naphthalenetrisulfonate, 1,3,6-naphthalenetrisulfonic acid, trisodium salt, naphthalene-1,3,6-trisulfonic acid trisodium salt, trisodium naphthalene-1,3,6-trisulfonate, sodium naphthalene-1,3,6-trisulfonate, trisodium 1,3,6-naphthalene trisulfonate, trisodium naphthalene-1,3,6-trisulphonate, 1,3,6-naphthalenetrisulfonic acid, sodium salt 1:3, 1,3,6-naphthalene trisulfonic acid trisodium salt, trisodium naphthalene trisulfonate
PubChem CID 78855
IUPAC Name trisodium naphthalene-1,3,6-trisulfonate
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Quantity 100 g
Formula Weight 434.29
Percent Purity ≥65.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Trisodium Naphthalene-1,3,6-trisulfonate Hydrate
EINECSNumber (4)-0471
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.