Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588984 Supplier Invitrogen™ Supplier No. PA588984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGEIVSADSM QVYEGLDIIT NKVSAQEQRI CRHHMISFVD PLVTNYTVVD FRNRATALIE DIFARDKIPI VVGGTNYYIE SLLWKVLVNT KPQEMGTEKV IDRKVELEKE DGLVLHKRLS QVDPEMAAKL HPHDKRKVAR SLQVFEETGI SHSEFLHRQH TEEGGGPLGG PLKFSNPCIL WLHADQAVLD ERLDKRVDDM LAAGLLEELR DFHRRYNQKN VSENSQDYQH GIFQSIGFKE FHEYLITEGK CTL; Positive Samples: HeLa; Cellular Location: Cytoplasm, Mitochondrion.

TRIT1 is responsible for the modification of A37 to isopentenyl A37 of both cytosolic and mitochondrial tRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Trit1
Gene Accession No. Q9H3H1
Gene Alias 2310075G14Rik; AI314189; AI314635; AV099619; GRO1; hGRO1; IPP transferase; IPPT; IPT; IPTase; isopentenyl-diphosphate:tRNA isopentenyltransferase; MOD5; Trit1; tRNA dimethylallyltransferase; tRNA dimethylallyltransferase, mitochondrial; tRNA isopentenylpyrophosphate transferase; tRNA isopentenyltransferase; tRNA isopentenyltransferase 1; tRNA isopentenyltransferase, mitochondrial
Gene Symbols Trit1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 48-300 of human TRIT1 (NP_0601162).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54802
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.