Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIT1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS17134R

Description
TRIT1 is responsible for the modification of A37 to isopentenyl A37 of both cytosolic and mitochondrial tRNAs.Specifications
TRIT1 | |
Polyclonal | |
Unconjugated | |
Trit1 | |
2310075G14Rik; AI314189; AI314635; AV099619; BOS_3767; GRO1; hGRO1; IPP transferase; IPPT; Ipt; IPTase; Isopentenyl-diphosphate:tRNA isopentenyltransferase; MOD5; TRIT1; tRNA dimethylallyltransferase; tRNA dimethylallyltransferase, mitochondrial; tRNA isopentenylpyrophosphate transferase; tRNA isopentenyltransferase; tRNA isopentenyltransferase 1; tRNA isopentenyltransferase, mitochondrial | |
Rabbit | |
Protein A | |
RUO | |
54802 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH | |
Q9H3H1 | |
Trit1 | |
KLH conjugated synthetic peptide derived from human TRIT1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction