Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIT1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | TRIT1 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TRIT1 Polyclonal antibody specifically detects TRIT1 in Human samples. It is validated for ImmunofluorescenceSpecifications
TRIT1 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
EC 2.5.1.75, EC 2.5.1.8, FLJ20061, hGRO1, IPP transferase, IPTase, IPTMGC149243, MGC149242, mitochondrial, MOD5, tRNA isopentenylpyrophosphate transferase, tRNA isopentenyltransferase, tRNA isopentenyltransferase 1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: KNVSENSQDYQHGIFQSIGFKEFHEYLITEGKCTLETSNQLLKKGIEALKQVTKRYARKQNRWVKNRFLSRPGPIVPPVYGLEVSDVSKWE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
54802 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title