Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRITC Polyclonal Antibody
GREENER_CHOICE

Catalog No. A6397
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A6397 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A6397 Supplier Invitrogen™ Supplier No. A6397
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

We offer a variety of anti-fluorescent dye antibodies that recognize specific fluorophores and, in most cases, quench their fluorescence. These anti-dye antibodies, including those that recognize the tetramethylrhodamine (TRITC) and Texas Red™ fluorophores, can also serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria, or other biomolecules have been internalized by endocytic or pinocytic processes. Our anti-tetramethylrhodamine (A6397) and anti-Texas Red (A6399) antibodies have been raised against the tetramethylrhodamine and Texas Red fluorophores, respectively. These antibodies quench much of the fluorescence of the complementary dyes. However, due to the related chemical structures, the anti-tetramethylrhodamine antibody crossreacts with the Texas Red fluorophore and the anti-Texas Red antibody cross-reacts with tetramethylrhodamine. Also, both antibodies tightly bind the Rhodamine Red™ fluorophore. Thus, either antibody can serve as an effective probe for tetramethylrhodamine, Texas Red, or Rhodamine Red dyes. We use a sensitive quenching assay to ensure that this antibody is provided at a consistently high titer value. As supplied, 20 μL of the antibody solution is certified to produce ≥50% of the maximal fluorescence quenching of 1 mL of a 50 nM solution of tetramethylrhodamine, assayed in 100 mM sodium phosphate, pH 8.0. Maximal quenching for tetramethylrhodamine is ~80% of the fluoresce...

Tetramethylrhodamine (TRITC) is a bright orange-fluorescent traditional dye with excitation ideally suited to the 532 nm laser line. It has been used extensively to label protein conjugates such as phalloidins for cytoskeleton counterstaining or antibody conjugates for immunofluorescence and other cellular imaging applications. Unlike biotin, which is an endogenous ligand in mitochondria, dye-based haptens permit background-free staining of cells and tissues and a sensitive alternative to biotin-streptavidin methods.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRITC
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 5mM sodium azide; pH 7.2
Gene Alias Tetramethylrhodamine isothiocyanate
Host Species Rabbit
Immunogen Tetramethylrhodamine.
Purification Method Purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.