Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Triton™X-100 Surfactant, OmniPur™, Calbiochem™,

Catalog No. 94404L Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
20 L
Packaging:
Glass Bottle
Plastic Receptacle in Fiberboard Box
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
94404L 4 L Plastic Receptacle in Fiberboard Box
94-001-00ML 100 mL Glass Bottle
94-101-L 1 L Glass Bottle
946020L 20 L Plastic Receptacle in Fiberboard Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 94404L Supplier MilliporeSigma™ Supplier No. 94404L

Used for solubilization of membrane proteins, enzymology, analytical and diagnostic work, as well as chromatography and electrophoresis

Recommended Storage

Ok to freeze

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Polyoxyethylene(10) Octylphenyl Ether
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Undesignated
Physical Form Viscous liquid
pH 6.0 to 8.0
Quantity 4 L
Packaging Plastic Receptacle in Fiberboard Box
Water <0.5%
Grade Molecular Biology
storageNote1 Ok to freeze
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.