Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triton(R) X-100, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle
Steel Can
Description
Spectrum™ Chemical Triton(R) X-100
Specifications
Specifications
CAS Max % | 1 |
Quantity | 20 L |
Packaging | Steel Can |
Grade | Reagent |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction