Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Triton™ X-100, 98%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. p-209823
Click to view available options
Quantity:
100 mL
250 mL
Packaging Type:
Glass bottle

CAS: 9002-93-1 | C16H26O2, C16H26O2 | 250.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For molecular biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Synonym Polyethylene glycol mono [4-(1,1,3,3-tetramethylbutyl)phenyl] ether

Specifications

Color Undesignated
Density 1.0670 g/mL
Quantity 100 mL
Formula Weight 646.85
MDL Number MFCD00132505
Melting Point 6.0°C
Packaging Type Glass bottle
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
pH 6 to 8 (5% aq. soln.)
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
Molecular Weight (g/mol) 250.38
Purity 98%, Molecular Biology
Solubility Solubility in water: soluble. Other solubilities: soluble in toluene, xylene, trichloroethylene,, ethylene glycol, ethyl ether, ethyl alcohol,, isopropyl alcohol, ethylene dichloride and most, other solvnets., insoluble in kerosene, miniral spirits.
Form Viscous Liquid
Assay 98%
Chemical Name or Material Triton™ X-100
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.