Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trityl Bromide 98.0+%, TCI America™
SDP

Catalog No. T0512100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0512100G 100 g
T051225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0512100G Supplier TCI America Supplier No. T0512100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 596-43-0
Molecular Formula C19H15Br
Molecular Weight (g/mol) 323.233
MDL Number MFCD00000120
InChI Key NZHXEWZGTQSYJM-UHFFFAOYSA-N
Synonym bromotriphenylmethane, triphenylmethyl bromide, trityl bromide, triphenylbromomethane, bromodiphenylmethyl benzene, methane, bromotriphenyl, benzene, 1,1',1-bromomethylidyne tris, triphenylmethylbromide, bromomethanetriyl tribenzene, 2-bromo-1,1,1-triphenylethane
PubChem CID 11692
IUPAC Name [bromo(diphenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Br

Specifications

Melting Point 156°C
Quantity 100 g
UN Number 3261
Formula Weight 323.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trityl Bromide
RTECSNumber PA5550000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.