Learn More
Invitrogen™ TRK fused gene Polyclonal Antibody

Description
Immunogen sequence homology: Opossum (88%), Chicken (82%).
Specifications
Specifications
| Antigen | TRK fused gene |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | TFG |
| Gene Accession No. | Q92734 |
| Gene Alias | AI173908; HMSNP; protein TFG; SPG57; TF6; TFG; Trk-fused gene; TRK-fused gene protein; TRKT3; TRKT3 oncogene |
| Gene Symbols | TFG |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.