Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRK fused gene Recombinant Rabbit Monoclonal Antibody (0V5P10)
GREENER_CHOICE

Catalog No. PIMA537850
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537850 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537850 Supplier Invitrogen™ Supplier No. MA537850
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Jurkat, Mouse testis, Mouse brain Immunogen sequence: APAFSGQPQQ LPAQPPQQYQ ASNYPAQTYT AQTSQPTNYT VAPASQPGMA PSQPGAYQPR PGFTSLPGST MTPPPSGPNP YARNRPPFGQ GYTQPGPGYR.

TFG protein, a recently identified novel protein, is a human homolog of mouse TFG protein. It contains an N-terminal coiled-coil structure, a SPYGQ-rich region followed by octicosapeptide repeats. Research shows that TFG can modulate SHP-1 activity and is a novel member of the NF-kB pathway. The physiological functions of this protein remain largely elusive, but TFG is thought to function by associating with other proteins, including TANK and NEMO. TFG was first identified as a partner of NTRK1 and is involved in generating the thyroid TRK-T3 oncogene, as well as oncogenic rearrangements with ALK in anaplastic lymphoma and NOR1 in mixoid chondrosarcoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRK fused gene
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 0V5P10
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TFG
Gene Accession No. Q92734
Gene Alias AI173908; HMSNP; protein TFG; SPG57; TF6; TFG; Trk-fused gene; TRK-fused gene protein; TRKT3; TRKT3 oncogene
Gene Symbols TFG
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 301-400 of human TRK fused gene (TFG) (Q92734).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10342, 21787, 360709
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.