Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkA Antibody (165131R), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1751RM
Description
TrkA Monoclonal antibody specifically detects TrkA in Human samples. It is validated for Flow Cytometry, CyTOF, Gene Knock-OutSpecifications
TrkA | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF, Gene Knock-Out | |
165131R | |
Flow Cytometry, CyTOF-ready, Knockout Validated | |
DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
Mouse myeloma cell line NS0-derived recombinant human TrkA, Ala33-Glu407, Accession # P04629 | |
0.1 mL | |
Adaptive Immunity, Cancer, Immunology, Neuroscience, Protein Kinase | |
4914 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction