Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkA Antibody (165131R), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1751RMFV450
Description
TrkA Monoclonal antibody specifically detects TrkA in Human samples. It is validated for Flow Cytometry, CyTOF, KnockDownSpecifications
| TrkA | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase | |
| 4914 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF, KnockDown | |
| 165131R | |
| Flow Cytometry, CyTOF-ready, Knockout Validated | |
| DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
| Mouse myeloma cell line NS0-derived recombinant human TrkA, Ala33-Glu407, Accession # P04629 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction