Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkA Antibody (MNAC13), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281074AF594
Description
TrkA Monoclonal specifically detects TrkA in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
TrkA | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
NTRK1 | |
Human TrkA ECD. | |
0.1 mL | |
Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase | |
4914 | |
Human | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
MNAC13 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody binds to the extracellular domain of TrkA. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction