Learn More
Promega TRKA Kinase Enzyme System, 1mg
Catalog No. PRV2932 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2932 1 mg
1 options
Catalog No. PRV2932 Supplier Promega Supplier No. V2932
Add to Cart
Add to Cart
Request a Quote
Recombinant human TRKA (amino acids 440-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TRKA is a member of the trk proto-oncogene family and encodes a 140kDa membrane-spanning protein tyrosine kinase.
  • Easily Screen and Profile TRKA Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C