Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TrkA Recombinant Rabbit Monoclonal Antibody (9F4Y9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535449
Description
Immunogen sequence: ESILYRKFTT ESDVWSFGVV LWEIFTYGKQ PWYQLSNTEA IDCITQGREL ERPRACPPEV YAIMRGCWQR EPQQRHSIKD VHARLQALAQ APPVYLDVLG.
NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.
Specifications
TrkA | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04629, P35739, Q3UFB7 | |
Ntrk1 | |
A synthetic peptide corresponding to a sequence within amino acids 697-796 of human TrkA (P04629). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
9F4Y9 | |
Unconjugated | |
Ntrk1 | |
C80751; DKFZp781I14186; EC 2.7.10.1; gp140trk; high affinity nerve growth factor receptor; kinase TrkA; MTC; neurotrophic receptor tyrosine kinase 1; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Ntrk1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; Tkr; TRK; TRK1; TRK1-transforming tyrosine kinase protein; Trka; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18211, 4914, 59109 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction