Learn More
Invitrogen™ TrkA Polyclonal Antibody

Description
Positive test controls include: HeLa, K-562, NIH/3T3, Mouse liver, Mouse heart, Rat liver. The target is usually found in the following locations: Cell membrane, Early endosome membrane, Late endosome membrane, Single-pass type I membrane protein. Immunogen sequence: MLRGGRRGQL GWHSWAAGPG SLLAWLILAS AGAAPCPDAC CPHGSSGLRC TRDGALDSLH HLPGAENLTE LYIENQQHLQ HLELRDLRGL GELRNLTIVK SGLRFVAPDA FHFTPRLSRL NLSFNALESL SWKTVQGLSL QELVLSGNPL HCSCALRWLQ RWEEEGLGGV PEQKLQCHGQ GPLAHMPNAS CGVPTLKVQV PNASVDVGDD VLLRCQVEGR GLEQAGWILT ELEQSATVMK SGGLPSLGLT LANVTSDLNR KNVTCWAEND VGRAEVSVQV.
Specifications
Specifications
| Antigen | TrkA |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Ntrk1 |
| Gene Accession No. | P04629, P35739, Q3UFB7 |
| Gene Alias | C80751; DKFZp781I14186; EC 2.7.10.1; gp140trk; high affinity nerve growth factor receptor; kinase TrkA; MTC; neurotrophic receptor tyrosine kinase 1; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Ntrk1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; Tkr; TRK; TRK1; TRK1-transforming tyrosine kinase protein; Trka; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A |
| Gene Symbols | Ntrk1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.