Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TrkA Recombinant Mouse Monoclonal Antibody (MNAC13)
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547963
Description
Specificity: This antibody binds to the extracellular domain of TrkA. It was shown not to cross react with TrkB. Moreover MNAC13 is specific to TrkA with negligible reactivity to TrkB and TrkC.
NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.
Specifications
TrkA | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P04629 | |
Ntrk1 | |
Human TrkA ECD. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Inhibition Assays, Immunohistochemistry, Surface Plasmon Resonance | |
MNAC13 | |
Unconjugated | |
Ntrk1 | |
C80751; DKFZp781I14186; EC 2.7.10.1; gp140trk; high affinity nerve growth factor receptor; kinase TrkA; MTC; neurotrophic receptor tyrosine kinase 1; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Ntrk1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; Tkr; TRK; TRK1; TRK1-transforming tyrosine kinase protein; Trka; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A | |
Mouse | |
Protein A | |
RUO | |
4914 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction