Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrkA/TrkB/TrkC Recombinant Rabbit Monoclonal Antibody (RM423)
GREENER_CHOICE

Catalog No. PIMA536218
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536218 Supplier Invitrogen™ Supplier No. MA536218
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MA5-36218 reacts to human TRK. This antibody may also react to mouse or rat TRK, as predicted by immunogen homology.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TrkA/TrkB/TrkC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone RM423
Concentration 0.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene NTRK1
Gene Accession No. P04629, Q16288, Q16620
Gene Alias BDNF/NT-3 growth factors receptor; BDNF-tropomyosine receptor kinase B; C80751; gp140trk; GP145-TrkB; GP145-TrkB/GP95-TrkB; high affinity nerve growth factor receptor; MTC; neural receptor protein-tyrosine kinase (trkB); neurotrophic receptor tyrosine kinase 1; neurotrophic receptor tyrosine kinase 2; Neurotrophic tyrosine kinase receptor type 1; Neurotrophic tyrosine kinase receptor type 2; neurotrophic tyrosine kinase, receptor, type 1; neurotrophic tyrosine kinase, receptor, type 2; neurotrophic tyrosine receptor kinase type 2; Ntrk1; NTRK2; Oncogene TRK; p140-TrkA; RATTRKB1; Slow nerve growth factor receptor; Tkr; Tkrb; trk; TRK1; TRK1-transforming tyrosine kinase protein; TRKA; trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; TRKB; trk-B; TrkB tyrosine kinase; TRKB1; tropomyosin-related kinase A; Tropomyosin-related kinase B; Tyrosine kinase receptor; Tyrosine kinase receptor A; tyrosine kinase receptor B
Gene Symbols NTRK1, Ntrk2, NTRK3
Host Species Rabbit
Immunogen A peptide corresponding to the C-terminus of human TRK.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4914, 4915, 4916
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.