Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP229660
Description
TrkB Polyclonal specifically detects TrkB in Rat samples. It is validated for Western Blot.Specifications
TrkB | |
Polyclonal | |
Western Blot 1:1000 | |
Q63604 | |
NTRK2 | |
Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH. Accession # Q63604 | |
Affinity purified | |
RUO | |
Primary | |
Specific for the ∼93 kDa TrkB protein in Western blots. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
Rabbit | |
93 kDa | |
0.1 mL | |
GPCR, Phospho Specific, Protein Kinase, Signal Transduction | |
4915 | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction