Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkB Antibody (SR2271), Novus Biologicals™

Rabbit Monoclonal Antibody
$295.74 - $408.03
Specifications
| Antigen | TrkB |
|---|---|
| Clone | SR2271 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
Description
TrkB Monoclonal antibody specifically detects TrkB in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| TrkB | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
| A synthesized peptide derived from human TrkB (Uniprot #: P15209) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| SR2271 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Neuroendocrinology, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 4915 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title