Learn More
Promega TRKB Kinase Enzyme System, 10microg
Catalog No. PRV4048 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4048 10 μg
1 options
Catalog No. PRV4048 Supplier Promega Supplier No. V4048
Recombinant human TRKB (455-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TRKB is a member of the neurotrophic tyrosine receptor kinase (NTRK) family.
  • Easily Screen and Profile TRKB Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C