Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkB, p Tyr707, p Tyr706 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $682.00
Specifications
Concentration | 1 mg/mL |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Target Species | Human, Mouse, Rat |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25476405
![]() |
Novus Biologicals
NBP2547640.05MG |
0.05 mg |
Each for $426.00
|
|
|||||
NBP254764
![]() |
Novus Biologicals
NBP254764 |
0.1 mg |
Each for $682.00
|
|
|||||
NB413816
![]() |
Novus Biologicals
NBP2547640.025MG |
0.025 mg |
Each for $196.10
|
|
|||||
Description
TrkB Polyclonal specifically detects TrkB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
1 mg/mL | |
Unconjugated | |
Human, Mouse, Rat | |
4915 | |
Trk B (phospho Tyr706/Tyr707) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 706/tyrosine 707 (T-D-Y (p)-Y (p)-R-V)derived from Human Trk B. | |
Primary |
Polyclonal | |
Rabbit | |
BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
NTRK2 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title