Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkB, p Tyr816 Antibody - Azide and BSA Free, Novus Biologicals™

Sheep Polyclonal Antibody
$158.60 - $478.00
Specifications
Antigen | TrkB (p Tyr816) |
---|---|
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TrkB Polyclonal specifically detects TrkB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TrkB (p Tyr816) | |
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry | |
Unconjugated | |
Sheep | |
Phospho Specific, Protein Kinase, Signal Transduction | |
Q16620 | |
4915 | |
IgG | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
NTRK2 | |
Primary | |
TrkB. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title