Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkC Antibody (101), Alexa Fluor™ 488

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290532AF488
Description
TrkC Monoclonal specifically detects TrkC in Mouse samples. It is validated for ELISA.Specifications
TrkC | |
Monoclonal | |
Alexa Fluor 488 | |
EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TrkC (NP_032772.3; Met1-Thr429). | |
0.1 mL | |
Cancer, Phospho Specific, Protein Kinase | |
4916 | |
Mouse | |
Purified |
ELISA | |
101 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Human TrkC | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction