Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540929 Supplier Invitrogen™ Supplier No. PA540929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NPGRFHTSER IRGVLSVITE ELPDVPLYYT LDQLSSTIHC NTPSLLQLRS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

TRMT1 is a tRNA(m(2)(2)G(26))dimethyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TRMT1
Gene Accession No. Q9NXH9
Gene Alias 6720406L13Rik; 6720477L24; D8Ertd812e; FLJ20244; im:7150454; LOC553298 protein; n(2),N(2)-dimethylguanosine tRNA methyltransferase; N(2),N(2)-dimethylguanosine tRNA methyltransferase; tRNA (guanine(26)-N(2))-dimethyltransferase; RGD1305992; TRM1; TRM1 tRNA methyltransferase 1; TRM1 tRNA methyltransferase 1 homolog; TRMT1; tRNA (guanine(26)-N(2))-dimethyltransferase; tRNA 2,2-dimethylguanosine-26 methyltransferase; tRNA methyltransferase 1; tRNA methyltransferase 1 homolog; tRNA methyltransferase 1 homolog (S. cerevisiae); tRNA(guanine-26,N(2)-N(2)) methyltransferase; tRNA(m(2,2)G26)dimethyltransferase
Gene Symbols TRMT1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human FLJ20244.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55621
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.