Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599786 Supplier Invitrogen™ Supplier No. PA599786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TRMT11.

TRMT11 belongs to the methyltransferase superfamily. It is a catalytic subunit of an S-adenosyl-L-methionine-dependent tRNA methyltransferase complex that mediates the methylation of the guanosine nucleotide at position 10 (m2G10) in tRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TRMT11
Gene Accession No. Q7Z4G4, Q9CWH5
Gene Alias 2410075D05Rik; 3110045I18Rik; AW213713; C6orf75; MDS024; putative RNA methylase MDS024; TRM11; Trmt11; TRMT11-1; tRNA (guanine(10)-N2)-methyltransferase homolog; tRNA guanosine-2'-O-methyltransferase TRM11 homolog; tRNA guanosine-2'-O-methyltransferase TRM11-like protein-like protein; tRNA methyltransferase 11; tRNA methyltransferase 11 homolog; tRNA methyltransferase 11 homolog (S. cerevisiae)
Gene Symbols TRMT11
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRMT11(Accession Q7Z4G4), corresponding to amino acid residues L394-E444.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60487, 73681
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.