Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT112 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120266 Supplier Invitrogen™ Supplier No. PA5120266
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, DU145, HeLa, Jurkat, Mouse brain, Mouse kidney, Mouse lung. Immunogen sequence: MKLLTHNLLS SHVRGVGSRG FPLRLQATEV RICPVEFNPN FVARMIPKVE WSAFLEAADN LRLIQVPKGP VEGYEENEEF LRTMHHLLLE VEVIEGTLQC PESGRMFPIS RGIPNMLLSE EETES.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT112
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene TRMT112
Gene Accession No. Q9DCG9, Q9UI30
Gene Alias AD-001; HSPC152; HSPC170; hTrm112; Multifunctional methyltransferase subunit TRM112-like protein; TRM112; TRM112-like protein; TRMT112; TRMT11-2; tRNA methyltransferase 112 homolog; tRNA methyltransferase 11-2 homolog (S. cerevisiae)
Gene Symbols TRMT112
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human TRMT112 (NP_057488.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293700, 51504, 67674
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.