Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT112 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558971 Supplier Invitrogen™ Supplier No. PA558971
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKLLTHNLLS SHVRGVGSRG FPLRLQATEV RICPVEFNPN FVARMIPKVE WSAFLEAADN LRL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT112
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRMT112
Gene Accession No. Q9UI30
Gene Alias AD-001; HSPC152; HSPC170; hTrm112; Multifunctional methyltransferase subunit TRM112-like protein; TRM112; TRM112-like protein; TRMT112; TRMT11-2; tRNA methyltransferase 112 homolog; tRNA methyltransferase 11-2 homolog (S. cerevisiae)
Gene Symbols TRMT112
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRMT112. Recombinant protein control fragment (Product #RP-97477).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51504
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.