Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555878 Supplier Invitrogen™ Supplier No. PA555878
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSVFERLQID IQHLCLNKIP VLREEKLPVV GIGKHLCGMA TDLALRCLVE TYAASFEERN EEPLAKRIKN DKTEKEIYTL AKEGNEKNVP EKWNPVAGIV IAL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

tRNA methylase which 2'-O-methylates cytidine(4) in tRNA(Pro) and tRNA(Gly)(GCC), and adenosine(4) in tRNA(His).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRMT13
Gene Accession No. Q9NUP7
Gene Alias 4631408H19Rik; A330067P21; A930028L21Rik; Ccdc76; coiled-coil domain containing 76; coiled-coil domain-containing protein 76; FLJ10287; FLJ11219; TRMT13; tRNA [Gm4] methyltransferase; tRNA guanosine-2'-O-methyltransferase TRM13 homolog; tRNA methyltransferase 13; tRNA methyltransferase 13 homolog; tRNA:m(4)X modification enzyme TRM13 homolog
Gene Symbols TRMT13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRMT13. Recombinant protein control fragment (Product #RP-94166).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54482
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.