Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555397 Supplier Invitrogen™ Supplier No. PA555397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QFSIHASSNV NKQEENGVFI KTTDDTTTDN YIAQGKRKSN EMITNLGKKQ KTDVSTEHPP FYYNIHRHSI KGMNMPKLKK FLCYLSQAGF RVSRTHFD Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.

This gene encodes a protein that has some similarity to N2,N2-dimethylguanosine tRNA methyltransferase from other organisms. Studies of the mouse ortholog have shown that this protein plays a role in motor coordination and exploratory behavior, and it may also be involved in modulating postnatal neuronal functions. Alternatively spliced transcripts have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT1L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Trmt1l
Gene Accession No. Q7Z2T5
Gene Alias 1190005F20Rik; AW226554; bG120K12.3; C1orf25; MST070; MSTP070; RGD1307890; TRM1 tRNA methyltransferase 1-like; TRM1L; Trm1-like; TRM1-like protein; TRMT1L; TRMT1-like protein; tRNA methyltransferase 1 homolog (S. cerevisiae)-like; tRNA methyltransferase 1 homolog-like; tRNA methyltransferase 1 like; tRNA methyltransferase 1-like
Gene Symbols Trmt1l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRMT1L. Recombinant protein control fragment (Product #RP-94154).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81627
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.